accessibility menu, dialog, popup

UserName

CH3H3CO

CAS RN 5779-93-1

IUPAC Name: 2,3-dimethylbenzaldehyde
Synonyms: 2,3-dimethylbenzaldehyde
Molecular Weight (g/mol): 134.18
Molecular Formula: C9H10O
InChi Key: UIFVCPMLQXKEEU-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C=O)=C1C
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)