missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethyl-2,3-dinitrobutane 98.0+%, TCI America™

$160.30 - $285.21

Chemical Identifiers

CAS 3964-18-9
Molecular Formula C6H12N2O4
Molecular Weight (g/mol) 176.172
MDL Number MFCD00007392
InChI Key DWCLXOREGBLXTD-UHFFFAOYSA-N
PubChem CID 77577
IUPAC Name 2,3-dimethyl-2,3-dinitrobutane
SMILES CC(C)(C(C)(C)[N+](=O)[O-])[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D182210G
View Documents
TCI America
D182210G
10 g
Each for $160.30
Only null left
 
D182225G
View Documents
TCI America
D182225G
25 g
Each for $285.21
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3964-18-9
176.172
DWCLXOREGBLXTD-UHFFFAOYSA-N
2,3-dimethyl-2,3-dinitrobutane
C6H12N2O4
MFCD00007392
77577
CC(C)(C(C)(C)[N+](=O)[O-])[N+](=O)[O-]

Specifications

3964-18-9
White
MFCD00007392
2926
CC(C)(C(C)(C)[N+](=O)[O-])[N+](=O)[O-]
176.172
176.17
Crystalline Powder
215°C
C6H12N2O4
10 g
DWCLXOREGBLXTD-UHFFFAOYSA-N
2,3-dimethyl-2,3-dinitrobutane
77577
≥98.0% (GC)
2,3-Dimethyl-2,3-dinitrobutane
Videos
Safety and Handling

Safety and Handling

RTECSNumber : EJ9943333

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'