CAS RN 3964-18-9
CAS RN 3964-18-9
IUPAC Name:
2,3-dimethyl-2,3-dinitrobutane
Synonyms:
2,3-dimethyl-2,3-dinitrobutane dmdnb
Molecular Weight (g/mol):
176.17
Molecular Formula:
C6H12N2O4
InChi Key:
DWCLXOREGBLXTD-UHFFFAOYSA-N
SMILES:
CC(C)([N+]([O-])=O)C(C)(C)[N+]([O-])=O
1
–
1
of
1
results
1
2,3-Dimethyl-2,3-dinitrobutane 98.0+%, TCI America™
CAS: 3964-18-9 Molecular Formula: C6H12N2O4 Molecular Weight (g/mol): 176.172 MDL Number: MFCD00007392 InChI Key: DWCLXOREGBLXTD-UHFFFAOYSA-N PubChem CID: 77577 IUPAC Name: 2,3-dimethyl-2,3-dinitrobutane SMILES: CC(C)(C(C)(C)[N+](=O)[O-])[N+](=O)[O-]