missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dihydroxynaphthalene 98.0+%, TCI America™
Supplier: TCI America D0593250G
Specifications
| 2,3-Dihydroxynaphthalene | |
| 163°C | |
| MFCD00004073 | |
| 2,3-dihydroxynaphthalene, 2,3-naphthalenediol, 2,3-dihydroxynapthalene, 2,3-dihydroxy naphthalene, 2,3-dihydroxynaphthlene, naphthalenediol-2,3, 3-hydroxy-2-naphthol, 2,3dihydroxynaphthalene, acmc-209rfr, 2,3-dihydroxylnaphthalene | |
| OC1=C(O)C=C2C=CC=CC2=C1 | |
| 160.17 | |
| CHEBI:38135 | |
| ≥98.0% (GC,T) |
| 92-44-4 | |
| C10H8O2 | |
| 250 g | |
| JRNGUTKWMSBIBF-UHFFFAOYSA-N | |
| naphthalene-2,3-diol | |
| 7091 | |
| 160.17 | |
| Crystalline Powder |
Chemical Identifiers
| 92-44-4 | |
| 160.17 | |
| JRNGUTKWMSBIBF-UHFFFAOYSA-N | |
| 7091 | |
| naphthalene-2,3-diol |
| C10H8O2 | |
| MFCD00004073 | |
| 2,3-dihydroxynaphthalene, 2,3-naphthalenediol, 2,3-dihydroxynapthalene, 2,3-dihydroxy naphthalene, 2,3-dihydroxynaphthlene, naphthalenediol-2,3, 3-hydroxy-2-naphthol, 2,3dihydroxynaphthalene, acmc-209rfr, 2,3-dihydroxylnaphthalene | |
| CHEBI:38135 | |
| OC1=C(O)C=C2C=CC=CC2=C1 |
Safety and Handling
EINECSNumber : (4)-0356
RTECSNumber : QJ4750000
TSCA : Yes