missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydroxynaphthalene 98.0+%, TCI America™

Catalog No. p-7126308
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D059325G 25 g
D0593250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D059325G Supplier TCI America Supplier No. D059325G
Only null left

Chemical Identifiers

CAS 92-44-4
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.17
MDL Number MFCD00004073
InChI Key JRNGUTKWMSBIBF-UHFFFAOYSA-N
Synonym 2,3-dihydroxynaphthalene, 2,3-naphthalenediol, 2,3-dihydroxynapthalene, 2,3-dihydroxy naphthalene, 2,3-dihydroxynaphthlene, naphthalenediol-2,3, 3-hydroxy-2-naphthol, 2,3dihydroxynaphthalene, acmc-209rfr, 2,3-dihydroxylnaphthalene
PubChem CID 7091
ChEBI CHEBI:38135
IUPAC Name naphthalene-2,3-diol
SMILES OC1=C(O)C=C2C=CC=CC2=C1

Specifications

Melting Point 163°C
Quantity 25 g
Formula Weight 160.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dihydroxynaphthalene
EINECSNumber (4)-0356
RTECSNumber QJ4750000
TSCA Yes