missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydroxybenzoic Acid 98.0+%, TCI America™

Catalog No. C00545G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C00545G 5 g
C005425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C00545G Supplier TCI America Supplier No. C00545G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 303-38-8
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.121
MDL Number MFCD00002446
InChI Key GLDQAMYCGOIJDV-UHFFFAOYSA-N
Synonym pyrocatechuic acid, o-pyrocatechuic acid, 2-pyrocatechuic acid, 3-hydroxysalicylic acid, dobk, dhba, 2,3-dihydroxybenzoicacid, benzoic acid, 2,3-dihydroxy, catecholcarboxylic acid, 2,3-dihydroxy-benzoic acid
PubChem CID 19
ChEBI CHEBI:18026
IUPAC Name 2,3-dihydroxybenzoic acid
SMILES C1=CC(=C(C(=C1)O)O)C(=O)O

Specifications

Melting Point 205°C
Quantity 5 g
Formula Weight 154.12
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dihydroxybenzoic Acid
EINECSNumber (3)-1616
RTECSNumber DG8576490
TSCA No