accessibility menu, dialog, popup

UserName

CAS RN 303-38-8

IUPAC Name: 2,3-dihydroxybenzoic acid
Synonyms: 2,3-dihydroxy-benzoic acid DOBK pyrocatechuic acid o-pyrocatechuic acid 2-pyrocatechuic acid
Molecular Weight (g/mol): 154.12
Molecular Formula: C7H6O4
InChi Key: GLDQAMYCGOIJDV-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(O)=C1O
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)