missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Difluorophenylacetic Acid 98.0+%, TCI America™
$240.64 - $240.64
Chemical Identifiers
| CAS | 145689-41-4 |
|---|---|
| Molecular Formula | C8H6F2O2 |
| Molecular Weight (g/mol) | 172.131 |
| MDL Number | MFCD00040968 |
| InChI Key | UXSQXUSJGPVOKT-UHFFFAOYSA-N |
| Synonym | 2,3-difluorophenylacetic acid, 2-2,3-difluorophenyl acetic acid, 2,3-difluorophenyl acetic acid, 2,3-difluorophenylaceticacid, benzeneacetic acid, 2,3-difluoro, 2,3-difluorobenzeneacetic acid, rarechem al bo 0630, pubchem1916, acmc-1crot |
| PubChem CID | 520772 |
| IUPAC Name | 2-(2,3-difluorophenyl)acetic acid |
| SMILES | C1=CC(=C(C(=C1)F)F)CC(=O)O |
Chemical Identifiers
| 145689-41-4 | |
| 172.131 | |
| UXSQXUSJGPVOKT-UHFFFAOYSA-N | |
| 520772 | |
| C1=CC(=C(C(=C1)F)F)CC(=O)O |
| C8H6F2O2 | |
| MFCD00040968 | |
| 2,3-difluorophenylacetic acid, 2-2,3-difluorophenyl acetic acid, 2,3-difluorophenyl acetic acid, 2,3-difluorophenylaceticacid, benzeneacetic acid, 2,3-difluoro, 2,3-difluorobenzeneacetic acid, rarechem al bo 0630, pubchem1916, acmc-1crot | |
| 2-(2,3-difluorophenyl)acetic acid |
Specifications
| 145689-41-4 | |
| White-Yellow | |
| MFCD00040968 | |
| 2,3-difluorophenylacetic acid, 2-2,3-difluorophenyl acetic acid, 2,3-difluorophenyl acetic acid, 2,3-difluorophenylaceticacid, benzeneacetic acid, 2,3-difluoro, 2,3-difluorobenzeneacetic acid, rarechem al bo 0630, pubchem1916, acmc-1crot | |
| C1=CC(=C(C(=C1)F)F)CC(=O)O | |
| 172.131 | |
| 172.13 | |
| Crystalline Powder |
| 118°C | |
| C8H6F2O2 | |
| 1 g | |
| UXSQXUSJGPVOKT-UHFFFAOYSA-N | |
| 2-(2,3-difluorophenyl)acetic acid | |
| 520772 | |
| ≥98.0% (GC,T) | |
| 2,3-Difluorophenylacetic Acid |
Safety and Handling
TSCA : No