accessibility menu, dialog, popup

UserName

OHOFF

CAS RN 145689-41-4

IUPAC Name: 2-(2,3-difluorophenyl)acetic acid
Synonyms: (2,3-difluorophenyl)acetic acid
Molecular Weight (g/mol): 172.13
Molecular Formula: C8H6F2O2
InChi Key: UXSQXUSJGPVOKT-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=CC(F)=C1F
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)