missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluorophenylacetic Acid 98.0+%, TCI America™

Catalog No. D38891G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D38891G Supplier TCI America Supplier No. D38891G

Chemical Identifiers

CAS 145689-41-4
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.131
MDL Number MFCD00040968
InChI Key UXSQXUSJGPVOKT-UHFFFAOYSA-N
Synonym 2,3-difluorophenylacetic acid, 2-2,3-difluorophenyl acetic acid, 2,3-difluorophenyl acetic acid, 2,3-difluorophenylaceticacid, benzeneacetic acid, 2,3-difluoro, 2,3-difluorobenzeneacetic acid, rarechem al bo 0630, pubchem1916, acmc-1crot
PubChem CID 520772
IUPAC Name 2-(2,3-difluorophenyl)acetic acid
SMILES C1=CC(=C(C(=C1)F)F)CC(=O)O

Specifications

Melting Point 118°C
Color White-Yellow
Quantity 1 g
Formula Weight 172.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Difluorophenylacetic Acid