missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluorophenylacetic acid, 98%

Catalog No. AAA1672803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1672803 1 g
AAA1672806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1672803 Supplier Thermo Scientific Chemicals Supplier No. A1672803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 145689-41-4 | C8H6F2O2 | 172.131 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 145689-41-4
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.131
MDL Number MFCD00040968
InChI Key UXSQXUSJGPVOKT-UHFFFAOYSA-N
Synonym 2,3-difluorophenylacetic acid, 2-2,3-difluorophenyl acetic acid, 2,3-difluorophenyl acetic acid, 2,3-difluorophenylaceticacid, benzeneacetic acid, 2,3-difluoro, 2,3-difluorobenzeneacetic acid, rarechem al bo 0630, pubchem1916, acmc-1crot
PubChem CID 520772
IUPAC Name 2-(2,3-difluorophenyl)acetic acid
SMILES C1=CC(=C(C(=C1)F)F)CC(=O)O

Specifications

Melting Point 117°C to 119°C
Quantity 1 g
Formula Weight 172.13
Percent Purity 98%
Chemical Name or Material 2,3-Difluorophenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only