missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Difluorobenzyl Bromide 98.0+%, TCI America™
$48.46 - $192.00
Chemical Identifiers
| CAS | 113211-94-2 |
|---|---|
| Molecular Formula | C7H5BrF2 |
| Molecular Weight (g/mol) | 207.018 |
| MDL Number | MFCD00042488 |
| InChI Key | FTBSGSZZESQDBM-UHFFFAOYSA-N |
| PubChem CID | 517984 |
| IUPAC Name | 1-(bromomethyl)-2,3-difluorobenzene |
| SMILES | C1=CC(=C(C(=C1)F)F)CBr |
Chemical Identifiers
| 113211-94-2 | |
| 207.018 | |
| FTBSGSZZESQDBM-UHFFFAOYSA-N | |
| 1-(bromomethyl)-2,3-difluorobenzene |
| C7H5BrF2 | |
| MFCD00042488 | |
| 517984 | |
| C1=CC(=C(C(=C1)F)F)CBr |
Specifications
| 113211-94-2 | |
| C7H5BrF2 | |
| 1 g | |
| 2,3-difluorobenzyl bromide, 1-bromomethyl-2,3-difluorobenzene, alpha-bromo-2,3-difluorotoluene, benzene, 1-bromomethyl-2,3-difluoro, 2,3-difluorobenzylbromide, a-bromo-2,3-difluorotoluene, 1-bromomethyl-2,3-difluoro-benzene, 2,3-difluorophenylbromide, 2, 3-difluorobenzyl bromide, pubchem4909 | |
| C1=CC(=C(C(=C1)F)F)CBr | |
| 207.018 | |
| 207.02 | |
| Liquid |
| Yellow | |
| MFCD00042488 | |
| 3265 | |
| FTBSGSZZESQDBM-UHFFFAOYSA-N | |
| 1-(bromomethyl)-2,3-difluorobenzene | |
| 517984 | |
| ≥98.0% (GC) | |
| 2,3-Difluorobenzyl Bromide |
Safety and Handling
TSCA : No