accessibility menu, dialog, popup

UserName

FBrF

CAS RN 113211-94-2

IUPAC Name: 1-(bromomethyl)-2,3-difluorobenzene
Synonyms: 1-(bromomethyl)-2,3-difluorobenzene
Molecular Weight (g/mol): 207.02
Molecular Formula: C7H5BrF2
InChi Key: FTBSGSZZESQDBM-UHFFFAOYSA-N
SMILES: FC1=CC=CC(CBr)=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results