missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluorobenzyl Bromide 98.0+%, TCI America™

Catalog No. D43205G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43205G 5 g
D43201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43205G Supplier TCI America Supplier No. D43205G

Chemical Identifiers

CAS 113211-94-2
Molecular Formula C7H5BrF2
Molecular Weight (g/mol) 207.018
MDL Number MFCD00042488
InChI Key FTBSGSZZESQDBM-UHFFFAOYSA-N
PubChem CID 517984
IUPAC Name 1-(bromomethyl)-2,3-difluorobenzene
SMILES C1=CC(=C(C(=C1)F)F)CBr

Specifications

Color Yellow
Quantity 5 g
UN Number 3265
Synonym 2,3-difluorobenzyl bromide, 1-bromomethyl-2,3-difluorobenzene, alpha-bromo-2,3-difluorotoluene, benzene, 1-bromomethyl-2,3-difluoro, 2,3-difluorobenzylbromide, a-bromo-2,3-difluorotoluene, 1-bromomethyl-2,3-difluoro-benzene, 2,3-difluorophenylbromide, 2, 3-difluorobenzyl bromide, pubchem4909
Formula Weight 207.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Difluorobenzyl Bromide