missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluorobenzoic Acid 98.0+%, TCI America™

Catalog No. p-7127089
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20585G 5 g
D205825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D20585G Supplier TCI America Supplier No. D20585G
Only null left

Chemical Identifiers

CAS 4519-39-5
Molecular Formula C7H3F2O2
Molecular Weight (g/mol) 157.10
MDL Number MFCD00010267
InChI Key JLZVIWSFUPLSOR-UHFFFAOYSA-M
Synonym 2,3-difluoro-benzoic acid, benzoic acid, 2,3-difluoro, difluorobenzoic acid, difluorobenzoic acid;, pubchem1379, 2,3 difluorobenzoic acid, acmc-209k0x, 2,3-difluoro benzoic acid, 2,3-difluorobenzoic acid;, ksc235o6t
PubChem CID 370590
IUPAC Name 2,3-difluorobenzoate
SMILES [O-]C(=O)C1=CC=CC(F)=C1F

Specifications

Melting Point 164°C
Color White
Quantity 5 g
Formula Weight 158.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Difluorobenzoic Acid