accessibility menu, dialog, popup

UserName

CAS RN 4519-39-5

IUPAC Name: 2,3-difluorobenzoate
Synonyms: 2,3-difluorobenzoate
Molecular Weight (g/mol): 157.1
Molecular Formula: C7H3F2O2
InChi Key: JLZVIWSFUPLSOR-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=CC(F)=C1F
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)