missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Difluoroaniline 97.0+%, TCI America™
$78.29 - $208.78
Chemical Identifiers
| CAS | 4519-40-8 |
|---|---|
| Molecular Formula | C6H5F2N |
| Molecular Weight (g/mol) | 129.11 |
| MDL Number | MFCD00010298 |
| InChI Key | YCCQGFYAVUTQFK-UHFFFAOYSA-N |
| Synonym | 2,3-difluorobenzenamine, 2,3-difluorophenylamine, benzenamine, 2,3-difluoro, 2,3-difluoro-phenylamine, difluoraniline, pubchem1530, acmc-1aiiw, 2,3-difluorobenzenamine?, 2,3-difluorophenyl amine, 2,3-difluoroaniline |
| PubChem CID | 78278 |
| IUPAC Name | 2,3-difluoroaniline |
| SMILES | C1=CC(=C(C(=C1)F)F)N |
Chemical Identifiers
| 4519-40-8 | |
| 129.11 | |
| YCCQGFYAVUTQFK-UHFFFAOYSA-N | |
| 78278 | |
| C1=CC(=C(C(=C1)F)F)N |
| C6H5F2N | |
| MFCD00010298 | |
| 2,3-difluorobenzenamine, 2,3-difluorophenylamine, benzenamine, 2,3-difluoro, 2,3-difluoro-phenylamine, difluoraniline, pubchem1530, acmc-1aiiw, 2,3-difluorobenzenamine?, 2,3-difluorophenyl amine, 2,3-difluoroaniline | |
| 2,3-difluoroaniline |
Specifications
| 4519-40-8 | |
| C6H5F2N | |
| 1 g | |
| 2,3-difluorobenzenamine, 2,3-difluorophenylamine, benzenamine, 2,3-difluoro, 2,3-difluoro-phenylamine, difluoraniline, pubchem1530, acmc-1aiiw, 2,3-difluorobenzenamine?, 2,3-difluorophenyl amine, 2,3-difluoroaniline | |
| C1=CC(=C(C(=C1)F)F)N | |
| 129.11 | |
| 129.11 | |
| Liquid |
| 69°C | |
| MFCD00010298 | |
| 2810 | |
| YCCQGFYAVUTQFK-UHFFFAOYSA-N | |
| 2,3-difluoroaniline | |
| 78278 | |
| ≥97.0% (GC) | |
| 2,3-Difluoroaniline |
Safety and Handling
TSCA : No