missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluoroaniline 97.0+%, TCI America™

Catalog No. D25951G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25951G 1 g
D25955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25951G Supplier TCI America Supplier No. D25951G
Only null left

Chemical Identifiers

CAS 4519-40-8
Molecular Formula C6H5F2N
Molecular Weight (g/mol) 129.11
MDL Number MFCD00010298
InChI Key YCCQGFYAVUTQFK-UHFFFAOYSA-N
Synonym 2,3-difluorobenzenamine, 2,3-difluorophenylamine, benzenamine, 2,3-difluoro, 2,3-difluoro-phenylamine, difluoraniline, pubchem1530, acmc-1aiiw, 2,3-difluorobenzenamine?, 2,3-difluorophenyl amine, 2,3-difluoroaniline
PubChem CID 78278
IUPAC Name 2,3-difluoroaniline
SMILES C1=CC(=C(C(=C1)F)F)N

Specifications

Boiling Point 69°C
Quantity 1 g
UN Number 2810
Formula Weight 129.11
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Difluoroaniline