missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluoro-6-nitrophenol 98.0+%, TCI America™

Catalog No. p-7127450
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27055G 5 g
D270525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27055G Supplier TCI America Supplier No. D27055G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 82419-26-9
Molecular Formula C6H3F2NO3
Molecular Weight (g/mol) 175.091
MDL Number MFCD00042255
InChI Key KEGOHDCHURMFKX-UHFFFAOYSA-N
Synonym 2,3-difluoro-6-nitrophenol, phenol, 2,3-difluoro-6-nitro, 2,3-difluoro-6-nitro-phenol, 6-nitro-2,3-difluorophenol, 2-hydroxy-3,4-difluoronitrobenzene, pubchem2873, acmc-209pot, 2,3 difluoro 6-nitrophenol, 2-nitro-5,6-difluorophenol, ksc495k9h
PubChem CID 2733745
SMILES C1=CC(=C(C(=C1[N+](=O)[O-])O)F)F

Specifications

Melting Point 63°C
Quantity 5 g
Formula Weight 175.09
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Difluoro-6-nitrophenol