missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluoro-6-methoxybenzoic acid, 97%

Catalog No. AAH2683009
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2683009 10 g
AAH2683003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2683009 Supplier Thermo Scientific Chemicals Supplier No. H2683009
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 773873-26-0 | C8H6F2O3 | 188.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 773873-26-0
Molecular Formula C8H6F2O3
Molecular Weight (g/mol) 188.13
MDL Number MFCD04116018
InChI Key MMTKYWQMCSZCGW-UHFFFAOYSA-N
Synonym 2,3-difluoro-6-methoxy-benzoic acid, pubchem4940, 2,3-difluoro-6-methoxybenzoicacid, benzoic acid,2,3-difluoro-6-methoxy
PubChem CID 3861323
IUPAC Name 2,3-difluoro-6-methoxybenzoic acid
SMILES COC1=C(C(=C(C=C1)F)F)C(=O)O

Specifications

Melting Point 126°C to 129°C
Quantity 10 g
Formula Weight 188.13
Percent Purity 97%
Chemical Name or Material 2,3-Difluoro-6-methoxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only