accessibility menu, dialog, popup

UserName

CAS RN 773873-26-0

IUPAC Name: 2,3-difluoro-6-methoxybenzoic acid
Synonyms: 2,3-difluoro-6-methoxybenzoic acid
Molecular Weight (g/mol): 188.13
Molecular Formula: C8H6F2O3
InChi Key: MMTKYWQMCSZCGW-UHFFFAOYSA-N
SMILES: COC1=CC=C(F)C(F)=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results