missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2',3'-Dideoxyinosine 98.0+%, TCI America™
$64.56 - $260.11
Chemical Identifiers
| CAS | 69655-05-6 |
|---|---|
| Molecular Formula | C10H12N4O3 |
| Molecular Weight (g/mol) | 236.23 |
| MDL Number | MFCD00077728 |
| InChI Key | BXZVVICBKDXVGW-JHPDDGAFNA-N |
| Synonym | didanosine, 2',3'-dideoxyinosine, dideoxyinosine, videx, videx ec, ddino, inosine, 2',3'-dideoxy, didanosina, didanosinum, ddi |
| PubChem CID | 50599 |
| ChEBI | CHEBI:490877 |
| IUPAC Name | 9-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one |
| SMILES | OC[C@@H]1CC[C@@H](O1)N1C=NC2=C1N=CNC2=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
D3066100MG
|
N/A | 100 mg |
Each for $64.56
|
|
|||||
|
D3066500MG
|
N/A | 500 mg |
Each for $260.11
|
|
|||||
Chemical Identifiers
| 69655-05-6 | |
| 236.23 | |
| BXZVVICBKDXVGW-JHPDDGAFNA-N | |
| 50599 | |
| 9-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one |
| C10H12N4O3 | |
| MFCD00077728 | |
| didanosine, 2',3'-dideoxyinosine, dideoxyinosine, videx, videx ec, ddino, inosine, 2',3'-dideoxy, didanosina, didanosinum, ddi | |
| CHEBI:490877 | |
| OC[C@@H]1CC[C@@H](O1)N1C=NC2=C1N=CNC2=O |
Specifications
| 69655-05-6 | |
| White | |
| MFCD00077728 | |
| BXZVVICBKDXVGW-JHPDDGAFNA-N | |
| 9-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one | |
| 50599 | |
| 236.23 | |
| Crystalline Powder |
| 182°C | |
| C10H12N4O3 | |
| didanosine, 2',3'-dideoxyinosine, dideoxyinosine, videx, videx ec, ddino, inosine, 2',3'-dideoxy, didanosina, didanosinum, ddi | |
| OC[C@@H]1CC[C@@H](O1)N1C=NC2=C1N=CNC2=O | |
| 236.23 | |
| CHEBI:490877 | |
| ≥98.0% (HPLC,T) | |
| 2′,3′-Dideoxyinosine |
Safety and Handling
RTECSNumber : NM7460700
TSCA : No