missing translation for 'onlineSavingsMsg'
Learn More

2',3'-Dideoxyinosine 98.0+%, TCI America™

Catalog No. D3066100MG
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3066100MG 100 mg
D3066500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3066100MG Supplier TCI America Supplier No. D3066100MG

Chemical Identifiers

CAS 69655-05-6
Molecular Formula C10H12N4O3
Molecular Weight (g/mol) 236.23
MDL Number MFCD00077728
InChI Key BXZVVICBKDXVGW-JHPDDGAFNA-N
Synonym didanosine, 2',3'-dideoxyinosine, dideoxyinosine, videx, videx ec, ddino, inosine, 2',3'-dideoxy, didanosina, didanosinum, ddi
PubChem CID 50599
ChEBI CHEBI:490877
IUPAC Name 9-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
SMILES OC[C@@H]1CC[C@@H](O1)N1C=NC2=C1N=CNC2=O

Specifications

Melting Point 182°C
Color White
Quantity 100 mg
Formula Weight 236.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′,3′-Dideoxyinosine
RTECSNumber NM7460700
TSCA No