accessibility menu, dialog, popup

UserName

CAS RN 69655-05-6

IUPAC Name: 9-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
Synonyms: 2'3'-dideoxyinosine
Molecular Weight (g/mol): 236.23
Molecular Formula: C10H12N4O3
InChi Key: BXZVVICBKDXVGW-JHPDDGAFNA-N
SMILES: OC[C@@H]1CC[C@@H](O1)N1C=NC2=C1N=CNC2=O
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)