missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dicyanohydroquinone, 98%

Catalog No. p-107238
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC232050050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC232050050 Supplier Thermo Scientific Chemicals Supplier No. 232050050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4733-50-0 | C8H4N2O2 | 160.132 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4733-50-0
MDL Number MFCD00001790
InChI Key MPAIWVOBMLSHQA-UHFFFAOYSA-N
Synonym 2,3-dicyanohydroquinone, 3,6-dihydroxyphthalonitrile, 2,3-dicyano-p-hydroquinone, 3,6-dihydroxyphthalodinitrile, 1,2-benzenedicarbonitrile, 3,6-dihydroxy, 2,3-dicyanobenzene-1,4-diol, phthalonitrile, 3,6-dihydroxy, dicyanohydroquinone, 2,3-dicyanohydroquinme, acmc-1asrw
PubChem CID 78467
IUPAC Name 3,6-dihydroxybenzene-1,2-dicarbonitrile
SMILES C1=CC(=C(C(=C1O)C#N)C#N)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >230.0°C
Color Brown to Khaki
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C8H4N2O2
Linear Formula (NC)2C6H2-1,4-(OH)2
Quantity 5 g
Solubility Information Solubility in water: soluble in hot water. Other solubilities: sparingly soluble in cold water,soluble in ether,acetone,ethanol,sparingly soluble in cold water,benzene,chlorofo,rm
Molecular Weight (g/mol) 160.13
Formula Weight 160.13
Percent Purity 98%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 2, 3-Dicyanohydroquinone, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic in contact with skin.
Causes skin irritation.
Toxic if inhaled.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fre
EINECSNumber 225-241-0
TSCA TSCA

RUO – Research Use Only