accessibility menu, dialog, popup

UserName

CAS RN 4733-50-0

IUPAC Name: 3,6-dihydroxybenzene-1,2-dicarbonitrile
Synonyms: 3,6-dihydroxybenzene-1,2-dicarbonitrile
Molecular Weight (g/mol): 160.13
Molecular Formula: C8H4N2O2
InChi Key: MPAIWVOBMLSHQA-UHFFFAOYSA-N
SMILES: OC1=CC=C(O)C(C#N)=C1C#N
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)