missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dichloropyridine 98.0+%, TCI America™
Supplier: TCI America D290625G
Specifications
| 2,3-Dichloropyridine | |
| 68°C | |
| C5H3Cl2N | |
| 25 g | |
| MAKFMOSBBNKPMS-UHFFFAOYSA-N | |
| 2,3-dichloropyridine | |
| 16988 | |
| ≥98.0% (GC) |
| 2402-77-9 | |
| White-Yellow | |
| MFCD00006229 | |
| 2,3-dichloro-pyridine, pyridine, 2,3-dichloro, dichloropyridine, pyridine, dichloro, ccris 1726, 2,3-dichloro pyridine, pubchem1198, pyridine,3-dichloro, 2.3-dichloro-pyridine, acmc-209g7x | |
| ClC1=CC=CN=C1Cl | |
| 147.99 | |
| 147.99 | |
| Crystalline Powder |
Chemical Identifiers
| 2402-77-9 | |
| 147.99 | |
| MAKFMOSBBNKPMS-UHFFFAOYSA-N | |
| 16988 | |
| ClC1=CC=CN=C1Cl |
| C5H3Cl2N | |
| MFCD00006229 | |
| 2,3-dichloro-pyridine, pyridine, 2,3-dichloro, dichloropyridine, pyridine, dichloro, ccris 1726, 2,3-dichloro pyridine, pubchem1198, pyridine,3-dichloro, 2.3-dichloro-pyridine, acmc-209g7x | |
| 2,3-dichloropyridine |
Safety and Handling
RTECSNumber : US8050000
TSCA : No