missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloropyridine 98.0+%, TCI America™

Catalog No. p-7127560
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
D29065G 5 g
D290625G 25 g
2 options

Catalog No. D29065G

Supplier: TCI America D29065G

Only null left

Chemical Identifiers

CAS 2402-77-9
Molecular Formula C5H3Cl2N
Molecular Weight (g/mol) 147.99
MDL Number MFCD00006229
InChI Key MAKFMOSBBNKPMS-UHFFFAOYSA-N
Synonym 2,3-dichloro-pyridine, pyridine, 2,3-dichloro, dichloropyridine, pyridine, dichloro, ccris 1726, 2,3-dichloro pyridine, pubchem1198, pyridine,3-dichloro, 2.3-dichloro-pyridine, acmc-209g7x
PubChem CID 16988
IUPAC Name 2,3-dichloropyridine
SMILES ClC1=CC=CN=C1Cl

Specifications

Melting Point 68°C
Color White-Yellow
Quantity 5 g
Formula Weight 147.99
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dichloropyridine
RTECSNumber US8050000
TSCA No