accessibility menu, dialog, popup

UserName

ClNCl

CAS RN 2402-77-9

IUPAC Name: 2,3-dichloropyridine
Synonyms: 2,3-dichloropyridine
Molecular Weight (g/mol): 147.99
Molecular Formula: C5H3Cl2N
InChi Key: MAKFMOSBBNKPMS-UHFFFAOYSA-N
SMILES: ClC1=CC=CN=C1Cl
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)