missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorophenylhydrazine Hydrochloride 97.0+%, TCI America™

Catalog No. p-7127419
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
D26415G 5 g
D264125G 25 g
2 options

Catalog No. D26415G

Supplier: TCI America D26415G

Only null left

Chemical Identifiers

CAS 21938-47-6
Molecular Formula C6H6Cl2N2
Molecular Weight (g/mol) 177.03
MDL Number MFCD00052265
InChI Key HPIALSXRVQRGMK-UHFFFAOYSA-N
Synonym 2,3-dichlorophenylhydrazine hydrochloride, 2,3-dichlorophenyl hydrazine hydrochloride, hydrazine, 2,3-dichlorophenyl-, monohydrochloride, 1-2,3-dichlorophenyl hydrazine hydrochloride, pubchem7561, acmc-209fpn, ksc494k0t, 2,3-dichlorophenylhydrazine, chloride, 2,3-dichlorophenylhydrazine hcl
PubChem CID 12276572
IUPAC Name (2,3-dichlorophenyl)hydrazine
SMILES NNC1=CC=CC(Cl)=C1Cl

Specifications

Quantity 5 g
Formula Weight 213.49
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dichlorophenylhydrazine Hydrochloride
EINECSNumber (3)-0472
TSCA No