accessibility menu, dialog, popup

UserName

H2NNHClCl

CAS RN 21938-47-6

IUPAC Name: (2,3-dichlorophenyl)hydrazine
Synonyms: (2,3-dichlorophenyl)hydrazine
Molecular Weight (g/mol): 177.03
Molecular Formula: C6H6Cl2N2
InChi Key: HPIALSXRVQRGMK-UHFFFAOYSA-N
SMILES: NNC1=CC=CC(Cl)=C1Cl
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)