missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorophenylhydrazine hydrochloride, 97%

Catalog No. p-7080953
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1071706 5 g
AAA1071714 25 g
AAA1071722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1071706 Supplier Thermo Scientific Chemicals Supplier No. A1071706
Only null left

CAS: 21938-47-6 | C6H6Cl2N2 | 177.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21938-47-6
Molecular Formula C6H6Cl2N2
Molecular Weight (g/mol) 177.03
MDL Number MFCD00052265
InChI Key HPIALSXRVQRGMK-UHFFFAOYSA-N
Synonym 2,3-dichlorophenylhydrazine hydrochloride, 2,3-dichlorophenyl hydrazine hydrochloride, hydrazine, 2,3-dichlorophenyl-, monohydrochloride, 1-2,3-dichlorophenyl hydrazine hydrochloride, pubchem7561, acmc-209fpn, ksc494k0t, 2,3-dichlorophenylhydrazine, chloride, 2,3-dichlorophenylhydrazine hcl
PubChem CID 12276572
IUPAC Name (2,3-dichlorophenyl)hydrazine;hydrochloride
SMILES NNC1=CC=CC(Cl)=C1Cl

Specifications

Melting Point ∼204°C (decomposition)
Quantity 5 g
Beilstein 3708910
Formula Weight 213.49
Percent Purity 97%
Chemical Name or Material 2,3-Dichlorophenylhydrazine hydrochloride
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only