Learn More
2,3-Dichlorophenyl isothiocyanate, 97%
CAS: 6590-97-2 | C7H3Cl2NS | 204.07 g/mol
$75.55 - $241.85
Chemical Identifiers
| CAS | 6590-97-2 |
|---|---|
| Molecular Formula | C7H3Cl2NS |
| Molecular Weight (g/mol) | 204.07 |
| MDL Number | MFCD00041048 |
| InChI Key | RQZIODPVCCTBAQ-UHFFFAOYSA-N |
| PubChem CID | 2796281 |
| IUPAC Name | 1,2-dichloro-3-isothiocyanatobenzene |
| SMILES | ClC1=CC=CC(N=C=S)=C1Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2093006
|
Thermo Scientific Chemicals
B2093006 |
5 g |
Each for $75.55
|
|
|||||
|
AAB2093014
|
Thermo Scientific Chemicals
B2093014 |
25 g |
Each for $241.85
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 6590-97-2 | |
| 204.07 | |
| RQZIODPVCCTBAQ-UHFFFAOYSA-N | |
| 1,2-dichloro-3-isothiocyanatobenzene |
| C7H3Cl2NS | |
| MFCD00041048 | |
| 2796281 | |
| ClC1=CC=CC(N=C=S)=C1Cl |
Specifications
| 6590-97-2 | |
| 256°C to 258°C | |
| Irritating | |
| 1.674 | |
| 5 g | |
| 2091585 | |
| 2,3-dichlorophenyl isothiocyanate, 2,3-dichlorophenylisothiocyanate, benzene,1,2-dichloro-3-isothiocyanato, benzene, 1,2-dichloro-3-isothiocyanato, dichlorophenylisothiocyanate, dichlorophenyl isothiocyanate, acmc-1b4g1, 2,3-dichlorophenyl-isothiocyanate, 1,2-dichloro-3-isothiocyanato-benzene, 2,3-dichloro-1-isothiocyanato-benzene | |
| ClC1=CC=CC(N=C=S)=C1Cl | |
| 204.07 | |
| 204.08 | |
| 2,3-Dichlorophenyl isothiocyanate |
| 1.434 | |
| >110°C (230°F) | |
| C7H3Cl2NS | |
| MFCD00041048 | |
| UN2922 | |
| Moisture sensitive | |
| RQZIODPVCCTBAQ-UHFFFAOYSA-N | |
| 1,2-dichloro-3-isothiocyanatobenzene | |
| 2796281 | |
| 97% |
Safety and Handling
GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only