missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorophenyl isothiocyanate, 97%

Catalog No. AAB2093006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2093006 5 g
AAB2093014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2093006 Supplier Thermo Scientific Chemicals Supplier No. B2093006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6590-97-2 | C7H3Cl2NS | 204.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6590-97-2
Molecular Formula C7H3Cl2NS
Molecular Weight (g/mol) 204.07
MDL Number MFCD00041048
InChI Key RQZIODPVCCTBAQ-UHFFFAOYSA-N
PubChem CID 2796281
IUPAC Name 1,2-dichloro-3-isothiocyanatobenzene
SMILES ClC1=CC=CC(N=C=S)=C1Cl

Specifications

Density 1.434
Boiling Point 256°C to 258°C
Flash Point >110°C (230°F)
Odor Irritating
Refractive Index 1.674
Quantity 5 g
UN Number UN2922
Beilstein 2091585
Sensitivity Moisture sensitive
Synonym 2,3-dichlorophenyl isothiocyanate, 2,3-dichlorophenylisothiocyanate, benzene,1,2-dichloro-3-isothiocyanato, benzene, 1,2-dichloro-3-isothiocyanato, dichlorophenylisothiocyanate, dichlorophenyl isothiocyanate, acmc-1b4g1, 2,3-dichlorophenyl-isothiocyanate, 1,2-dichloro-3-isothiocyanato-benzene, 2,3-dichloro-1-isothiocyanato-benzene
Formula Weight 204.08
Percent Purity 97%
Chemical Name or Material 2,3-Dichlorophenyl isothiocyanate
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only