missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorophenol 99.0+%, TCI America™

Catalog No. D039125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D039125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D039125G Supplier TCI America Supplier No. D039125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 576-24-9
Molecular Formula C6H4Cl2O
Molecular Weight (g/mol) 163.00
MDL Number MFCD00002160
InChI Key UMPSXRYVXUPCOS-UHFFFAOYSA-N
Synonym phenol, 2,3-dichloro, dichlorophenol, unii-izm39u444l, ccris 5902, 2,3-dichloro phenol, 2,3 dichlorophenol, phenol,3-dichloro, pubchem3696, 2,3-dichloro-phenol, acmc-1ar0l
PubChem CID 11334
IUPAC Name 2,3-dichlorophenol
SMILES OC1=CC=CC(Cl)=C1Cl

Specifications

Melting Point 59°C
Boiling Point 90°C
Quantity 25 g
UN Number 2020
Formula Weight 163.00
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dichlorophenol
EINECSNumber (3)-0930
RTECSNumber SK8450000
TSCA Yes