accessibility menu, dialog, popup

UserName

OHClCl

CAS RN 576-24-9

IUPAC Name: 2,3-dichlorophenol
Synonyms: 2,3-dichlorophenol
Molecular Weight (g/mol): 163
Molecular Formula: C6H4Cl2O
InChi Key: UMPSXRYVXUPCOS-UHFFFAOYSA-N
SMILES: OC1=CC=CC(Cl)=C1Cl
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)