missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorophenol, 98+%

CAS: 576-24-9 | C6H4Cl2O | 163.00 g/mol

$58.62 - $481.93

Chemical Identifiers

CAS 576-24-9
Molecular Formula C6H4Cl2O
Molecular Weight (g/mol) 163.00
MDL Number MFCD00002160
InChI Key UMPSXRYVXUPCOS-UHFFFAOYSA-N
Synonym phenol, 2,3-dichloro, dichlorophenol, unii-izm39u444l, ccris 5902, 2,3-dichloro phenol, 2,3 dichlorophenol, phenol,3-dichloro, pubchem3696, 2,3-dichloro-phenol, acmc-1ar0l
PubChem CID 11334
IUPAC Name 2,3-dichlorophenol
SMILES OC1=CC=CC(Cl)=C1Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2294314
View Documents
Thermo Scientific Chemicals
B2294314
25 g
Each for $58.62
Only null left
 
AAB2294322
View Documents
Thermo Scientific Chemicals
B2294322
100 g
Each for $149.00
Only null left
 
AAB2294336
View Documents
Thermo Scientific Chemicals
B2294336
500 g
Each for $481.93
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

576-24-9
163.00
UMPSXRYVXUPCOS-UHFFFAOYSA-N
11334
OC1=CC=CC(Cl)=C1Cl
C6H4Cl2O
MFCD00002160
phenol, 2,3-dichloro, dichlorophenol, unii-izm39u444l, ccris 5902, 2,3-dichloro phenol, 2,3 dichlorophenol, phenol,3-dichloro, pubchem3696, 2,3-dichloro-phenol, acmc-1ar0l
2,3-dichlorophenol

Specifications

576-24-9
206°C
Phenol-like
MFCD00002160
UN2020
phenol, 2,3-dichloro, dichlorophenol, unii-izm39u444l, ccris 5902, 2,3-dichloro phenol, 2,3 dichlorophenol, phenol,3-dichloro, pubchem3696, 2,3-dichloro-phenol, acmc-1ar0l
OC1=CC=CC(Cl)=C1Cl
163.00
163
2,3-Dichlorophenol
57°C to 60°C
115°C (239°F)
C6H4Cl2O
25 g
2043615
UMPSXRYVXUPCOS-UHFFFAOYSA-N
2,3-dichlorophenol
11334
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 209-399-8

RTECSNumber : SK8450000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only