missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dichloronitrobenzene 99.0+%, TCI America™
$54.06 - $369.08
Chemical Identifiers
| CAS | 3209-22-1 |
|---|---|
| Molecular Formula | C6H3Cl2NO2 |
| Molecular Weight (g/mol) | 191.995 |
| MDL Number | MFCD00007062 |
| InChI Key | CMVQZRLQEOAYSW-UHFFFAOYSA-N |
| Synonym | 2,3-dichloronitrobenzene, 2,3-dichloro-1-nitrobenzene, benzene, 1,2-dichloro-3-nitro, benzene, dichloronitro, unii-985ews35kg, ccris 5901, pubchem10770, acmc-1ccaw, 2,3 dichloronitrobenzene, dichloronitrobenzene |
| PubChem CID | 18555 |
| IUPAC Name | 1,2-dichloro-3-nitrobenzene |
| SMILES | C1=CC(=C(C(=C1)Cl)Cl)[N+](=O)[O-] |
Chemical Identifiers
| 3209-22-1 | |
| 191.995 | |
| CMVQZRLQEOAYSW-UHFFFAOYSA-N | |
| 18555 | |
| C1=CC(=C(C(=C1)Cl)Cl)[N+](=O)[O-] |
| C6H3Cl2NO2 | |
| MFCD00007062 | |
| 2,3-dichloronitrobenzene, 2,3-dichloro-1-nitrobenzene, benzene, 1,2-dichloro-3-nitro, benzene, dichloronitro, unii-985ews35kg, ccris 5901, pubchem10770, acmc-1ccaw, 2,3 dichloronitrobenzene, dichloronitrobenzene | |
| 1,2-dichloro-3-nitrobenzene |
Specifications
| 3209-22-1 | |
| 257°C | |
| MFCD00007062 | |
| 2811 | |
| CMVQZRLQEOAYSW-UHFFFAOYSA-N | |
| 1,2-dichloro-3-nitrobenzene | |
| 18555 | |
| ≥99.0% (GC) | |
| 2,3-Dichloronitrobenzene |
| 62°C | |
| C6H3Cl2NO2 | |
| 25 g | |
| 2,3-dichloronitrobenzene, 2,3-dichloro-1-nitrobenzene, benzene, 1,2-dichloro-3-nitro, benzene, dichloronitro, unii-985ews35kg, ccris 5901, pubchem10770, acmc-1ccaw, 2,3 dichloronitrobenzene, dichloronitrobenzene | |
| C1=CC(=C(C(=C1)Cl)Cl)[N+](=O)[O-] | |
| 191.995 | |
| 192.00 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-0455
RTECSNumber : CZ5240000
TSCA : Yes