CAS RN 3209-22-1
CAS RN 3209-22-1
IUPAC Name:
1,2-dichloro-3-nitrobenzene
Synonyms:
2,3-dichloronitrobenzene
Molecular Weight (g/mol):
192
Molecular Formula:
C6H3Cl2NO2
InChi Key:
CMVQZRLQEOAYSW-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=CC(Cl)=C1Cl
1
–
1
of
1
results
1
2,3-Dichloronitrobenzene 99.0+%, TCI America™
CAS: 3209-22-1 Molecular Formula: C6H3Cl2NO2 Molecular Weight (g/mol): 191.995 MDL Number: MFCD00007062 InChI Key: CMVQZRLQEOAYSW-UHFFFAOYSA-N Synonym: 2,3-dichloronitrobenzene,2,3-dichloro-1-nitrobenzene,benzene, 1,2-dichloro-3-nitro,benzene, dichloronitro,unii-985ews35kg,ccris 5901,pubchem10770,acmc-1ccaw,2,3 dichloronitrobenzene,dichloronitrobenzene PubChem CID: 18555 IUPAC Name: 1,2-dichloro-3-nitrobenzene SMILES: C1=CC(=C(C(=C1)Cl)Cl)[N+](=O)[O-]