missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dichlorobenzyl Cyanide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 70°C |
| Color | White-Yellow |
| Boiling Point | 115°C |
| Quantity | 5 g |
| UN Number | 3439 |
| Formula Weight | 186.04 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,3-Dichlorobenzyl Cyanide |
Safety and Handling
| TSCA | No |