CAS RN 3218-45-9
IUPAC Name:
2-(2,3-dichlorophenyl)acetonitrile
Synonyms:
2-(2,3-dichlorophenyl)acetonitrile
Molecular Weight (g/mol):
186.04
Molecular Formula:
C8H5Cl2N
InChi Key:
CZLINJDTCHACEH-UHFFFAOYSA-N
SMILES:
ClC1=CC=CC(CC#N)=C1Cl
1
–
1
of
1
results
1
2,3-Dichlorobenzyl Cyanide 98.0+%, TCI America™
CAS: 3218-45-9 Molecular Formula: C8H5Cl2N Molecular Weight (g/mol): 186.04 MDL Number: MFCD01861392 InChI Key: CZLINJDTCHACEH-UHFFFAOYSA-N Synonym: 2,3-Dichlorophenylacetonitrile PubChem CID: 229020 IUPAC Name: 2-(2,3-dichlorophenyl)acetonitrile SMILES: ClC1=CC=CC(CC#N)=C1Cl