missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dichlorobenzyl Alcohol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 86°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 177.02 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,3-Dichlorobenzyl Alcohol |
Safety and Handling
| TSCA | No |