accessibility menu, dialog, popup

UserName

HOClCl

CAS RN 38594-42-2

IUPAC Name: (2,3-dichlorophenyl)methanol
Synonyms: (2,3-dichlorophenyl)methanol
Molecular Weight (g/mol): 177.02
Molecular Formula: C7H6Cl2O
InChi Key: STVBVTWXWZMRPZ-UHFFFAOYSA-N
SMILES: OCC1=CC=CC(Cl)=C1Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results