missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dichloro-5-nitro-1,4-naphthoquinone 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 272.04 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,3-Dichloro-5-nitro-1,4-naphthoquinone |
Safety and Handling
| TSCA | No |