accessibility menu, dialog, popup

UserName

NOOOOClCl

CAS RN 22360-86-7

IUPAC Name: 2,3-dichloro-5-nitro-1,4-dihydronaphthalene-1,4-dione
Synonyms: 2,3-dichloro-5-nitronaphthalene-1,4-dione
Molecular Weight (g/mol): 272.04
Molecular Formula: C10H3Cl2NO4
InChi Key: AWCITCQRUBWCJA-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC2=C1C(=O)C(Cl)=C(Cl)C2=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results