missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloro-1,4-naphthoquinone 95.0+%, TCI America™

Catalog No. p-7126177
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D038425G 25 g
D0384500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D038425G Supplier TCI America Supplier No. D038425G
Only null left

Chemical Identifiers

CAS 117-80-6
Molecular Formula C10H4Cl2O2
Molecular Weight (g/mol) 227.04
MDL Number MFCD00001677
InChI Key SVPKNMBRVBMTLB-UHFFFAOYSA-N
Synonym 2,3-dichloro-1,4-naphthoquinone, dichlone, diclone, 2,3-dichloronaphthoquinone, phygon, algistat, sanquinon, uniroyal, phygon paste, dichloronaphthoquinone
PubChem CID 8342
IUPAC Name 2,3-dichloro-1,4-dihydronaphthalene-1,4-dione
SMILES ClC1=C(Cl)C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 196°C
Color Yellow
Quantity 25 g
UN Number 2811
Formula Weight 227.04
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dichloro-1,4-naphthoquinone
EINECSNumber (4)-0378
RTECSNumber QL7525000
TSCA Yes