Learn More
2,3-Dichloro-1,4-naphthoquinone, 98%
CAS: 117-80-6 | C10H4Cl2O2 | 227.04 g/mol
Supplier: Thermo Scientific Chemicals A1337836
Description
Use in textiles, as organic catalyst, as chemical intermediate, as additive in dye binders. Also used as an important catalyst to produce 3,3-dichlorobenzene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUse as seed disinfectant, textiles, as organic catalyst, as chemical intermediate, as additive in dye binders, as antifouling agent. Also used to control blue-green algae in ponds, lakes, and swimming pools. Also used as an important catalyst to produce 3,3-Dichloro benzine.
Solubility
Soluble in water (0.008g/L). Soluble in hot toluene almost transparency.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
| 2,3-Dichloro-1,4-naphthoquinone | |
| 193°C to 196°C (sublimation) | |
| C10H4Cl2O2 | |
| 500 g | |
| 1073511 | |
| 2,3-dichloro-1,4-naphthoquinone, dichlone, diclone, 2,3-dichloronaphthoquinone, phygon, algistat, sanquinon, uniroyal, phygon paste, dichloronaphthoquinone | |
| SVPKNMBRVBMTLB-UHFFFAOYSA-N | |
| 2,3-dichloro-1,4-dihydronaphthalene-1,4-dione | |
| 8342 | |
| 98% |
| 117-80-6 | |
| 275°C (2 mmHg) | |
| MFCD00001677 | |
| UN2811 | |
| 14,3045 | |
| Soluble in water (0.008g/L). Soluble in hot toluene almost transparency. | |
| ClC1=C(Cl)C(=O)C2=CC=CC=C2C1=O | |
| 227.04 | |
| 227.05 |
Chemical Identifiers
| 117-80-6 | |
| 227.04 | |
| SVPKNMBRVBMTLB-UHFFFAOYSA-N | |
| 8342 |
| C10H4Cl2O2 | |
| MFCD00001677 | |
| 2,3-dichloro-1,4-naphthoquinone, dichlone, diclone, 2,3-dichloronaphthoquinone, phygon, algistat, sanquinon, uniroyal, phygon paste, dichloronaphthoquinone | |
| ClC1=C(Cl)C(=O)C2=CC=CC=C2C1=O |
Safety and Handling
GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
H302-H315-H319-H500
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 204-210-5
RTECSNumber : QL7525000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only