missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromobutane 99.0+%, TCI America™

Catalog No. p-7126073
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D017725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D017725G Supplier TCI America Supplier No. D017725G

Chemical Identifiers

CAS 5408-86-6
Molecular Formula C4H8Br2
Molecular Weight (g/mol) 215.916
MDL Number MFCD00000146
InChI Key BXXWFOGWXLJPPA-UHFFFAOYSA-N
Synonym butane, 2,3-dibromo, dl-2,3-dibromobutane, +/--2,3-dibromobutane, ss-butylene bromide, .+-.-2,3-dibromobutane, dibromo butane, butane,3-dibromo, beta-butylene bromide, r*,r*-+--2,3-dibromobutane, pubchem12534
PubChem CID 21508
IUPAC Name 2,3-dibromobutane
SMILES CC(C(C)Br)Br

Specifications

Boiling Point 157°C
Quantity 25 g
Formula Weight 215.92
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Dibromobutane