missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dibromo-5-chloropyridine 98.0+%, TCI America™
Supplier: TCI America D438125G
Specifications
| 2,3-Dibromo-5-chloropyridine | |
| 43°C | |
| MFCD08687254 | |
| 2,3 dibromo-5-chloro pyridine, 5-chloro-2,3-dibromopyridine, 2,3-dibromo-5-chloro-pyridine, pyridine, 2,3-dibromo-5-chloro, pubchem5397, acmc-209cej, ksc494o0p, 2,3-dibromo-5-chloro pyridine, 2,3-dibromo-5-dichloropyridine, abbypharma ap-30-4531 | |
| ClC1=CC(Br)=C(Br)N=C1 | |
| 271.34 | |
| 271.34 | |
| Crystalline Powder |
| 137628-17-2 | |
| C5H2Br2ClN | |
| 25 g | |
| GDUFWKJMOOVEMX-UHFFFAOYSA-N | |
| 2,3-dibromo-5-chloropyridine | |
| 11000162 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 137628-17-2 | |
| 271.34 | |
| GDUFWKJMOOVEMX-UHFFFAOYSA-N | |
| 11000162 | |
| ClC1=CC(Br)=C(Br)N=C1 |
| C5H2Br2ClN | |
| MFCD08687254 | |
| 2,3 dibromo-5-chloro pyridine, 5-chloro-2,3-dibromopyridine, 2,3-dibromo-5-chloro-pyridine, pyridine, 2,3-dibromo-5-chloro, pubchem5397, acmc-209cej, ksc494o0p, 2,3-dibromo-5-chloro pyridine, 2,3-dibromo-5-dichloropyridine, abbypharma ap-30-4531 | |
| 2,3-dibromo-5-chloropyridine |
Safety and Handling
TSCA : No