missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromo-5-chloropyridine 98.0+%, TCI America™

Catalog No. p-7128250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43815G 5 g
D438125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43815G Supplier TCI America Supplier No. D43815G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 137628-17-2 | C5H2Br2ClN | 271.34 g/mol

Chemical Identifiers

CAS 137628-17-2
Molecular Formula C5H2Br2ClN
Molecular Weight (g/mol) 271.34
MDL Number MFCD08687254
InChI Key GDUFWKJMOOVEMX-UHFFFAOYSA-N
Synonym 2,3 dibromo-5-chloro pyridine, 5-chloro-2,3-dibromopyridine, 2,3-dibromo-5-chloro-pyridine, pyridine, 2,3-dibromo-5-chloro, pubchem5397, acmc-209cej, ksc494o0p, 2,3-dibromo-5-chloro pyridine, 2,3-dibromo-5-dichloropyridine, abbypharma ap-30-4531
PubChem CID 11000162
IUPAC Name 2,3-dibromo-5-chloropyridine
SMILES ClC1=CC(Br)=C(Br)N=C1

Specifications

Melting Point 43°C
Quantity 5 g
Formula Weight 271.34
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dibromo-5-chloropyridine